missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NENF Protein

A cDNA sequence encoding the NENF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12829-1mg

 View more versions of this product

Product Code. 15961679

  • £5730.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NENF Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASGQTPRPAERG PPVRLFTEEE LARYGGEEED QPIYLAVKGV VFDVTSGKEF YGRGAPYNAL TGKDSTRGVA KMSLDPADLT HDTTGLTAKE LEALDEVFTK VYKAKYPIVG YTARRILNED GSPNLDFKPE DQPHFDIKDE F
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NENF was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers