missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NHP2-like protein 1
A DNA sequence encoding the Homo sapiens (Human) NHP2-like protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9307-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003796.1 | |
NHP2-like protein 1 | |
Research Use Only | |
NHP2L1 | |
Recombinant Protein | |
Yeast | |
TEADVNPKAY PLADAHLTKK LLDLVQQSCN YKQLRKGANE ATKTLNRGIS EFIVMAADAE PLEIILHLPL LCEDKNVPYV FVRSKQALGR ACGVSRPVIA CSVTIKEGSQ LKQQIQSIQQ SIERLLV | |
Greater than 90% as determined by SDS-PAGE. |
4809 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |