missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NK-tumor recognition Protein
A DNA sequence encoding the Homo sapiens (Human) NK-tumor recognition protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6422-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_005376.2 | |
NK-tumor recognition Protein | |
Research Use Only | |
NKTR | |
Recombinant Protein | |
E. coli | |
HFDIEINREP VGRIMFQLFS DICPKTCKNF LCLCSGEKGL GKTTGKKLCY KGSTFHRVVK NFMIQGGDFS EGNGKGGESI YGGYFKDENF ILKHDRAFLL SMANRGKHTN GSQFFITTKP APHLDGVHVV FGLVISGFEV IEQIENLKTD AASRPYADVR VIDCGV | |
Greater than 90% as determined by SDS-PAGE. |
4820 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |