missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NKIRAS1 Protein
A cDNA sequence encoding the NKIRAS1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12848-1ug
This item is not returnable.
View return policy
Specifications
NKIRAS1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGKGCKVVVC GLLSVGKTAI LEQLLYGNHT IGMEDCETME DVYMASVETD RGVKEQLHLY DTRGLQEGVE LPKHYFSFAD GFVLVYSVNN LESFQRVELL KKEIDKFKDK KEVAIVVLGN KIDLSEQRQV DAEVAQQWAK SEKVRLWEVT VTDRKTLIEP FTLLASKLSQ PQSKSSFPLP GRKNKGNSNS EN | |
Greater than 95.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NKIRAS1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |