missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NME2 Protein
A cDNA sequence encoding the NME2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12855-25ug
This item is not returnable.
View return policy
Specifications
NME2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANLERTFIA IKPDGVQRGL VGEIIKRFEQ KGFRLVAMKF LRASEEHLKQ HYIDLKDRPF FPGLVKYMNS GPVVAMVWEG LNVVKTGRVM LGETNPADSK PGTIRGDFCI QVGRNIIHGS DSVKSAEKEI SLWFKPEELV DYKSCAHDWV YE | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The NME2 protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol. |