missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleoredoxin-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Nucleoredoxin-like protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7712-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001155097.1 | |
Nucleoredoxin-like protein 1 | |
Research Use Only | |
NXNL2 | |
Recombinant Protein | |
E. coli | |
MVDILGERHL VTCKGATVEA EAALQNKVVA LYFAAARCAP SRDFTPLLCD FYTALVAEAR RPAPFEVVFV SADGSCQEML DFMRELHGAW LALPFHDPYR QRSLALLPRL ECSGVILAHC NLCLLGSSDS LALAS | |
Greater than 90% as determined by SDS-PAGE. |
158046 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |