missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OMG Protein

A cDNA sequence encoding the OMG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12924-10ug

 View more versions of this product

Product Code. 15931969

  • £155.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

OMG Protein
Research Use Only
Recombinant Protein
Baculovirus
ICPLQCICTE RHRHVDCSGR NLSTLPSGLQ ENIIHLNLSY NHFTDLHNQL TQYTNLRTLD ISNNRLESLP AHLPRSLWNM SAANNNIKLL DKSDTAYQWN LKYLDVSKNM LEKVVLIKNT LRSLEVLNLS SNKLWTVPTN MPSKLHIVDL SNNSLTQILP GTLINLTNLT HLYLHNNKFT FIPDQSFDQL FQLQEITLYN NRWSCDHKQN ITYLLKWMME TKAHVIGTPC STQISSLKEH NMYPTPSGFT SSLFTVSGMQ TVDTINSLSV VTQPKVTKIP KQYRTKETTF GATLSKDTTF TSTDKAFVPY PEDTSTETIN SHEAAAATLT IHLQDGMVTN TSLTSSTKSS PTPMTLSITS GMPNNFSEMP QQSTTLNLWR EETTTNVKTP LPSVEHHHHH H
Greater than 95.0% as determined by analysis by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
OMG protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers