missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OSCAR Protein

A cDNA sequence encoding the OSCAR was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12933-1mg

 View more versions of this product

Product Code. 15931999

  • £2975.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

OSCAR Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDITPSVA IIVPPASYHP KPWLGAQPAT VVTPGVNVTL RCRAPQPAWR FGLFKPGEIA PLLFRDVSSE LAEFFLEEVT PAQGGIYRCC YRRPDWGPGV WSQPSDVLEL LVTEELPRPS LVALPGPVVG PGANVSLRCA GRLRNMSFVL YREGVAAPLQ YRHSAQPWAD FTLLGARAPG TYSCYYHTPS APYVLSQRSE VLVISWEGEG PEARPASSAP GMQAPGPPPS DPGAQAPSLS SFRPRGLVLQ PLLPQTQDSW DPAPPPSDPG V
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
OSCAR protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers