missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OSCAR Protein
A cDNA sequence encoding the OSCAR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12933-5ug
This item is not returnable.
View return policy
Specifications
OSCAR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDITPSVA IIVPPASYHP KPWLGAQPAT VVTPGVNVTL RCRAPQPAWR FGLFKPGEIA PLLFRDVSSE LAEFFLEEVT PAQGGIYRCC YRRPDWGPGV WSQPSDVLEL LVTEELPRPS LVALPGPVVG PGANVSLRCA GRLRNMSFVL YREGVAAPLQ YRHSAQPWAD FTLLGARAPG TYSCYYHTPS APYVLSQRSE VLVISWEGEG PEARPASSAP GMQAPGPPPS DPGAQAPSLS SFRPRGLVLQ PLLPQTQDSW DPAPPPSDPG V | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
OSCAR protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol. |