missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Osteoprotegerin/TNFRSF11B Protein
Recombinant Protein
£101.51 - £1147.92
Specifications
Accession Number | O00300, O00300, O00300 |
---|---|
For Use With (Application) | PAGE, Bioactivity |
Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB,150 mM NaCl, pH 6.0. |
Gene ID (Entrez) | 4982 |
Molecular Weight (g/mol) | M.W. (Theoretical): 19.7 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18488781
|
Novus Biologicals™
NBP2-35108-500ug |
500 μg |
£639.29
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18404902
|
Novus Biologicals™
NBP2-35108-100ug |
100 μg |
£305.61
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18431562
|
Novus Biologicals™
NBP2-35108-10ug |
10 μg |
£101.51
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18460342
|
Novus Biologicals™
NBP2-35108-1mg |
1mg |
£1147.92
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteoprotegerin Protein is fully biologically active when compared to standard. The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL).Specifications
O00300, O00300, O00300 | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB,150 mM NaCl, pH 6.0. | |
M.W. (Theoretical): 19.7 kDa | |
Less than 1 EU/μg of Osteoprotegerin/TNFRSF11B as determined by LAL method. | |
TNFRSF11B | |
Apoptosis, Biologically Active Proteins | |
Osteoprotegerin/TNFRSF11B |
PAGE, Bioactivity | |
4982 | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C | |
>95%, by SDS-PAGE and HPLC |