missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human OX40/TNFRSF4 Mouse IgG2a Fc Protein, CF
Carrier-Free Recombinant Protein
Brand: R&D Systems™ 10649-OX-050
336.93 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human OX40 Ligand/TNFSF4 Protein (Catalog # 1054-OX ) is immobilized at 250 ng/mL, the concentration of Recombinant Human OX40/TNFRSF4 Mouse IgG2a Fc Chimera that produces 50% optimal binding response is found to be approximately 2.5-15 ng/mL.Specifications
NP_003318.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 60-70 kDa, under reducing conditions, M.W. (Theoretical): 47 kDa | |
Mouse myeloma cell line, NS0-derived human OX40/TNFRSF4 protein Human OX40/TNFRSF4 (Leu29-Ala216) (N-terminus) Accession # NP_003318.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Unconjugated | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
3586 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
ACT-135, CD134, CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, Ly-70, TGIF, TNFRSF4, Txgp1, TXGP1L | |
Reconstitute at 250 μg/mL in PBS. | |
IL-10 |