missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PEBP1 Protein
A cDNA sequence encoding the PEBP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13001-1mg
This item is not returnable.
View return policy
Specifications
PEBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPVDLSKWSG PLSLQEVDEQ PQHPLHVTYA GAAVDELGKV LTPTQVKNRP TSISWDGLDS GKLYTLVLTD PDAPSRKDPK YREWHHFLVV NMKGNDISSG TVLSDYVGSG PPKGTGLHRY VWLVYEQDRP LKCDEPILSN RSGDHRGKFK VASFRKKYEL RAPVAGTCYQ AEWDDYVPKL YEQLSGK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PEBP1 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |