missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PEDF Protein
A cDNA sequence encoding the PEDF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13005-HIS-5ug
This item is not returnable.
View return policy
Specifications
5176 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
QNPASPPEEG SPDPDSTGAL VEEEDPFFKV PVNKLAAAVS NFGYDLYRVR SSTSPTTNVL LSPLSVATAL SALSLGAEQR TESIIHRALY YDLISSPDIH GTYKELLDTV TAPQKNLKSA SRIVFEKKLR IKSSFVAPLE KSYGTRPRVL TGNPRLDLQE INNWVQAQMK GKLARSTKEI PDEISILLLG VAHFKGQWVT KFDSRKTSLE DFYLDEERTV RVPMMSDPKA VLRYGLDSDL SCKIAQLPLT GSMSIIFFLP LKVTQNLTLI EESLTSEFIH DIDRELKTVQ AVLTVPKLKL SYEGEVTKSL QEMKLQSLFD SPDFSKITGK PIKLTQVEHR AGFEWNEDGA GTTPSPGLQP AHLTFPLDYH LNQPFIFVLR DTDTGALLFI GKILDPRGPA AADYKDDDDK | |
Greater than 95% as determined by SDS-PAGE. |
PEDF Protein | |
Research Use Only | |
SERPINF1 | |
Human | |
His | |
The filtered (0.4microm) concentrated (0.5 mg/ml) protein solution was lyophilized with 20mM Tris and 20mM NaCl pH 7.5. |