missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PFDN6 Protein

A cDNA sequence encoding the PFDN6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13017-10ug

 View more versions of this product

Product Code. 15932259

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

PFDN6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAELIQKKLQ GEVEKYQQLQ KDLSKSMSGR QKLEAQLTEN NIVKEELALL DGSNVVFKLL GPVLVKQELG EARATVGKRL DYITAEIKRY ESQLRDLERQ SEQQRETLAQ LQQEFQRAQA AKAGAPGKA
Greater than 95% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PFDN6 protein solution (0.5 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers