missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGK1 / PGKA Protein
A DNA sequence encoding the Homo sapiens (Human) PGK1 / PGKA, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000282.1 |
---|---|
Gene ID (Entrez) | 5230 |
Name | PGK1 / PGKA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053901
|
enQuireBio™
QP8305-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073901
|
enQuireBio™
QP8305-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063901
|
enQuireBio™
QP8305-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083901
|
enQuireBio™
QP8305-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093901
|
enQuireBio™
QP8305-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003911
|
enQuireBio™
QP8305-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000282.1 | |
PGK1 / PGKA Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5230 | |
Research Use Only | |
PGK1 | |
Recombinant Protein | |
E. coli | |
FNVPMKNNQI TNNQRIKAAV PSIKFCLDNG AKSVVLMSHL GRPDGVPMPD KYSLEPVAVE LKSLLGKDVL FLKDCVGPEV EKACANPAAG SVILLENLRF HVEEEGKGKD ASGNKVKAEP AKIEAFRASL SKLGDVYVND AFGTAHRAHS SMVGVNLPQK AGGFLMKKEL NYFAKALESP ERPFLAILGG AKVADKIQLI NNMLDKVNEM IIGGGMAFTF LKVLNNMEIG TSLFDEEGAK IVKDLMSKAE KNGVKITLPV DFVTADKFDE NAKTGQATVA SGIPAGWMGL DCGPESSKKY AEAVTRAKQI VWNGPVGVFE WEAFARGTKA LMDEVVKATS RGCITIIGGG DTATCCAKWN TEDKVSHVST GGGASLELLE GKVLPGVDAL SNI | |
Greater than 90% as determined by SDS-PAGE. |