missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGLS Protein
A cDNA sequence encoding the PGLS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13032-2ug
This item is not returnable.
View return policy
Specifications
PGLS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAPAPGLIS VFSSSQELGA ALAQLVAQRA ACCLAGARAR FALGLSGGSL VSMLARELPA AVAPAGPASL ARWTLGFCDE RLVPFDHAES TYGLYRTHLL SRLPIPESQV ITINPELPVE EAAEDYAKKL RQAFQGDSIP VFDLLILGVG PDGHTCSLFP DHPLLQEREK IVAPISDSPK PPPQRVTLTL PVLNAARTVI FVATGEGKAA VLKRILEDQE ENPLPAALVQ PHTGKLCWFL DEAAARLLTV PFEKHSTL | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PGLS protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 100mM NaCl and 10% glycerol. |