missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phylloquinone omega-hydroxylase CYP4F11 Protein
A DNA sequence encoding the Homo sapiens (Human) Phylloquinone omega-hydroxylase CYP4F11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8115-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001122404.1 | |
Phylloquinone omega-hydroxylase CYP4F11 Protein | |
Research Use Only | |
CYP4F11 | |
Recombinant Protein | |
E. coli | |
TYTFYDNCRR LQCFPQPPKQ NWFWGHQGLV TPTEEGMKTL TQLVTTYPQG FKLWLGPTFP LLILCHPDII RPITSASAAV APKDMIFYGF LKPWLGDGLL LSGGDKWSRH RRMLTPAFHF NILKPYMKIF NKSVNIMHDK WQRLASEGSA RLDMFEHISL MTLDSLQKCV FSFESNCQEK PSEYIAAILE LSAFVEKRNQ QILLHTDFLY YLTPDGQRFR RACHLVHDFT DAVIQERRCT LPTQGIDDFL KNKAKSKTLD FIDVLLLSKD EDGKELSDED IRAEADTFMF EGHDTTASGL SWVLYHLAKH PEYQEQCRQE VQELLKDREP IEIEWDDLAQ LPFLTMCIKE SLRLHPPVPV ISRCCTQDFV LPDGRVIPKG IVCLINIIGI HYNPTVWPDP EVYDPFRFDQ ENIKERSPLA FIPFSAGPRN CIGQAFAMAE MKVVLALTLL HFRILPTHTE PRRKPELILR AEGGLWLRVE PLGANSQ | |
Greater than 90% as determined by SDS-PAGE. |
57834 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |