missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PISD Protein
A DNA sequence encoding the Homo sapiens (Human) PISD, was expressed in the hosts and tags indicated.
£145.46 - £1840.00
Specifications
Accession Number | NP_001313340.1 |
---|---|
Gene ID (Entrez) | 23761 |
Name | PISD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082981
|
enQuireBio™
QP8162-ec-10ug |
10 μg | N/A | ||||||
16022991
|
enQuireBio™
QP8162-ec-50ug |
50 μg | N/A | ||||||
16072981
|
enQuireBio™
QP8162-ec-100ug |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16002991
|
enQuireBio™
QP8162-ec-200ug |
200 μg | N/A | ||||||
16012991
|
enQuireBio™
QP8162-ec-500ug |
500 μg | N/A | ||||||
16092981
|
enQuireBio™
QP8162-ec-1mg |
1 mg |
£1840.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001313340.1 | |
PISD Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
23761 | |
Research Use Only | |
PISD | |
Recombinant Protein | |
E. coli | |
MMCQSEARQG PELRAAKWLH FPQLALRRRL GQLSCMSRPA LKLRSWPLTV LYYLLPFGAL RPLSRVGWRP VSRVALYKSV PTRLLSRAWG RLNQVELPHW LRRPVYSLYI WTFGVNMKEA AVEDLHHYRN LSEFFRRKLK PQARPVCGLH SVISPSDGRI LNFGQVKNCE VEQVKGVTYS LESFLGPRMC TEDLPFPPAA SCDSFKNQLV TREGNELYHC VIYLAPGDYH CFHSPTDWTV SHRRHFPGSL MSVNPGMARW IKELFCHNER VVLTGDWKHG FFSLTAVGAT NVGSIRIYFD RDLHTNSPRH SKGSYNDFSF VTHTNREGVP MRKGEHLGEF NLGSTIVLIF EAPKDFNFQL KTGQKIRFGE ALGSL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human PISD Protein