missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Plasma Kallikrein/KLKB1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 2497-SE-010

210.79 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18300068

  • £276.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave a flourogenic peptide substrate Pro-Phe-Arg-7-amido-4-methylcoumarin (PFR-AMC). The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P03952
Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl.
M.W. (Observed): 80 kDa, reducing conditions; M.W. (theoretical): 70 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Enzyme Activity
3818
Plasma Kallikrein/KLKB1
Mouse myeloma cell line, NS0-derived human Plasma Kallikrein/KLKB1 protein Gly20-Ala638, with a C-terminal 6-His tag
Pro and processed forms
KLKB1
Recombinant
Plasma Kallikrein/KLKB1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers