missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plastin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Plastin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6289-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002289.2 | |
Plastin-2 Protein | |
Research Use Only | |
LCP1 | |
Recombinant Protein | |
Yeast | |
ARGSVSDEEM MELREAFAKV DTDGNGYISF NELNDLFKAA CLPLPGYRVR EITENLMATG DLDQDGRISF DEFIKIFHGL KSTDVAKTFR KAINKKEGIC AIGGTSEQSS VGTQHSYSEE EKYAFVNWIN KALENDPDCR HVIPMNPNTN DLFNAVGDGI VLCKMINLSV PDTIDERTIN KKKLTPFTIQ ENLNLALNSA SAIGCHVVNI GAEDLKEGKP YLVLGLLWQV IKIGLFADIE LSRNEALIAL LREGESLEDL MKLSPEELLL RWANYHLENA GCNKIGNFST DIKDSKAYYH LLEQVAPKGD EEGVPAVVID MSGLREKDDI QRAECMLQQA ERLGCRQFVT ATDVVRGNPK LNLAFIANLF NRYPALHKPE NQDIDWGALE GETREERTFR NWMNSLGVNP RVNHLYSDLS DALVIFQLYE KIKVPVDWNR VNKPPYPKLG GNMKKLENCN YAVELGKNQA KFSLVGIGGQ DLNEGNRTLT LALIWQLMRR YTLNILEEIG GGQKVNDDII VNWVNETLRE AKKSSSISSF KDPKISTSLP VLDLIDAIQP GSINYDLLKT ENLNDDEKLN NAKYAISMAR KIGARVYALP EDLVEVNPKM VMTVFACLMG KGMKRV | |
Greater than 90% as determined by SDS-PAGE. |
3936 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |