missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLSCR3 Protein
A cDNA sequence encoding the PLSCR3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13078-1mg
This item is not returnable.
View return policy
Specifications
PLSCR3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGYLPP KGYAPSPPPP YPVTPGYPEP ALHPGPGQAP VPAQVPAPAP GFALFPSPGP VALGSAAPFL PLPGVPSGLE FLVQIDQILI HQKAERVETF LGWETCNRYE LRSGAGQPLG QAAEESNCCA RLCCGARRPL RVRLADPGDR EVLRLLRPLH CGCSCCPCGL QEMEVQAPPG TTIGHVLQTW HPFLPKFSIQ DADRQTVLRV VGPCWTCGCG TDTNFEVKTR DESRSVGRIS KQWGGLVREA LTDADDFGLQ FPLDLDVR | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PLSCR3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M urea. |