missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLG2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) POLG2, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8185-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_009146.2 | |
POLG2, mitochondrial Protein | |
Research Use Only | |
POLG2 | |
Recombinant Protein | |
E. coli | |
MRSRVAVRAC HKVCRCLLSG FGGRVDAGQP ELLTERSSPK GGHVKSHAEL EGNGEHPEAP GSGEGSEALL EICQRRHFLS GSKQQLSRDS LLSGCHPGFG PLGVELRKNL AAEWWTSVVV FREQVFPVDA LHHKPGPLLP GDSAFRLVSA ETLREILQDK ELSKEQLVAF LENVLKTSGK LRENLLHGAL EHYVNCLDLV NKRLPYGLAQ IGVCFHPVFD TKQIRNGVKS IGEKTEASLV WFTPPRTSNQ WLDFWLRHRL QWWRKFAMSP SNFSSSDCQD EEGRKGNKLY YNFPWGKELI ETLWNLGDHE LLHMYPGNVS KLHGRDGRKN VVPCVLSVNG DLDRGMLAYL YDSFQLTENS FTRKKNLHRK VLKLHPCLAP IKVALDVGRG PTLELRQVCQ GLFNELLENG ISVWPGYLET MQSSLEQLYS KYDEMSILFT VLVTETTLEN GLIHLRSRDT TMKEMMHISK LKDFLIKYIS SAKNV | |
Greater than 90% as determined by SDS-PAGE. |
11232 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |