missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2E Protein
A cDNA sequence encoding the POLR2E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13098-20ug
This item is not returnable.
View return policy
Specifications
POLR2E Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDEEET YRLWKIRKTI MQLCHDRGYL VTQDELDQTL EEFKAQFGDK PSEGRPRRTD LTVLVAHNDD PTDQMFVFFP EEPKVGIKTI KVYCQRMQEE NITRALIVVQ QGMTPSAKQS LVDMAPKYIL EQFLQQELLI NITEHELVPE HVVMTKEEVTELLARYKLRE NQLPRIQAGD PVARYFGIKR GQVVKIIRPS ETAGRYITYR LVQ | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLR2E solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |