missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Polycomb complex protein BMI-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Polycomb complex protein BMI-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8441-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001190991.1 | |
Polycomb complex protein BMI-1 Protein | |
Research Use Only | |
BMI1 | |
Recombinant Protein | |
E. coli | |
MHRTTRIKIT ELNPHLMCVL CGGYFIDATT IIECLHSFCK TCIVRYLETS KYCPICDVQV HKTRPLLNIR SDKTLQDIVY KLVPGLFKNE MKRRRDFYAA HPSADAANGS NEDRGEVADE DKRIITDDEI ISLSIEFFDQ NRLDRKVNKD KEKSKEEVND KRYLRCPAAM TVMHLRKFLR SKMDIPNTFQ IDVMYEEEPL KDYYTLMDIA YIYTWRRNGP LPLKYRVRPT CKRMKISHQR DGLTNAGELE | |
Greater than 90% as determined by SDS-PAGE. |
100532731 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |