missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PQBP1 Protein
A cDNA sequence encoding the PQBP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13131-5ug
This item is not returnable.
View return policy
Specifications
PQBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMPLPVA LQTRLAKRGI LKHLEPEPEE EIIAEDYDDD PVDYEATRLE GLPPSWYKVF DPSCGLPYYW NADTDLVSWL SPHDPNSVVT KSAKKLRSSN ADAEEKLDRS HDKSDRGHDK SDRSHEKLDR GHDKSDRGHD KSDRDRERGY DKVDRERERD RERDRDRGYD KADREEGKER RHHRREELAP YPKSKKAVSR KDEELDPMDP SSYSDAPRGT WSTGLPKRNE AKTGADTTAA GPLFQQRPYP SPGAVLRANA EASRTKQQD | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PQBP1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl and 20% glycerol. |