missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable ATP-dependent RNA helicase DDX27 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable ATP-dependent RNA helicase DDX27, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7872-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060365.7 | |
Probable ATP-dependent RNA helicase DDX27 Protein | |
Research Use Only | |
DDX27 | |
Recombinant Protein | |
E. coli | |
DNMLADLGLI GTIGEDDEVP VEPESDSGDE EEEGPIVLGR RQKALGKNRS ADFNPDFVFT EKEGTYDGSW ALADVMSQLK KKRAATTLDE KIEKVRKKRK TEDKEAKSGK LEKEKEAKEG SEPKEQEDLQ ENDEEGSEDE ASETDYSSAD ENILTKADTL KVKDRKKKKK KGQEAGGFFE DASQYDENLS FQDMNLSRPL LKAITAMGFK QPTPIQKACI PVGLLGKDIC ACAATGTGKT AAFALPVLER LIYKPRQAPV TRVLVLVPTR ELGIQVHSVT RQLAQFCNIT TCLAVGGLDV KSQEAALRAA PDILIATPGR LIDHLHNCPS FHLSSIEVLI LDEADRMLDE YFEEQMKEII RMCSHHRQTM LFSATMTDEV KDLASVSLKN PVRIFVNSNT DVAPFLRQEF IRIRPNREGD REAIVAALLT RTFTDHVMLF TQTKKQAHRM HILLGLMGLQ VGELHGNLSQ TQRLEALRRF KDEQIDILVA TDVAARGLDI EGVKTVINFT MPNTIKHYVH RVGRTARAGR AGRSVSLVGE DERKMLKEIV KAAKAPVKAR ILPQDVILKF RDKIEKMEKD VYAVLQLEAE EKEMQQSEAQ INTAKRLLEK GKEAVVQEPE RSWFQTKEER KKEKIAKALQ EFDLALRGKK KRKKFMKDAK KKGEMTAEER SQFEILKAQM FAERLAKRNR RAKRARAMPE EEPVRGPAKK QKQGKKSVFD EELTNTSKKA LKQYRAGPSF EERKQLGLPH QRRGGNFKSK SRYKR | |
Greater than 90% as determined by SDS-PAGE. |
55661 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |