missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Procollagen C-endopeptidase enhancer 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Procollagen C-endopeptidase enhancer 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6472-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_037495.1 | |
Procollagen C-endopeptidase enhancer 2 Protein | |
Research Use Only | |
PCOLCE2 | |
Recombinant Protein | |
E. coli | |
QSPERPVFTC GGILTGESGF IGSEGFPGVY PPNSKCTWKI TVPEGKVVVL NFRFIDLESD NLCRYDFVDV YNGHANGQRI GRFCGTFRPG ALVSSGNKMM VQMISDANTA GNGFMAMFSA AEPNERGDQY CGGLLDRPSG SFKTPNWPDR DYPAGVTCVW HIVAPKNQLI ELKFEKFDVE RDNYCRYDYV AVFNGGEVND ARRIGKYCGD SPPAPIVSER NELLIQFLSD LSLTADGFIG HYIFRPKKLP TTTEQPVTTT FPVTTGLKPT VALCQQKCRR TGTLEGNYCS SDFVLAGTVI TTITRDGSLH ATVSIINIYK EGNLAIQQAG KNMSARLTVV CKQCPLLRRG LNYIIMGQVG EDGRGKIMPN SFIMMFKTKN QKLLDALKNK QC | |
Greater than 90% as determined by SDS-PAGE. |
26577 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |