missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Proinsulin C-Peptide Analogue Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-35211-10ug
This item is not returnable.
View return policy
Specifications
P01308, P01308, P01308 | |
Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. | |
M.W. (Theoretical): 3.6 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
INS | |
Use in WB reported in secitific publication PMID: 32433667 | |
Proinsulin C-Peptide Analogue |
Western Blot, PAGE | |
3630 | |
10 μg | |
Less than 0.1 EU/μg of Proinsulin C-Peptide Analogue as determined by LAL method. | |
Reconstitute with sterilized distilled water or 0.1% BSA aqueous buffer to a final concentration of 0.1 - 1.0 mg/ml. | |
>95%, by SDS-PAGE and HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction