missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein BEX3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein BEX3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6419-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_055195.1 | |
Protein BEX3 Protein | |
Research Use Only | |
NGFRAP1 | |
Recombinant Protein | |
E. coli | |
MANIHQENEE MEQPMQNGEE DRPLGGGEGH QPAGNRRGQA RRLAPNFRWA IPNRQINDGM GGDGDDMEIF MEEMREIRRK LRELQLRNCL RILMGELSNH HDHHDEFCLM P | |
Greater than 90% as determined by SDS-PAGE. |
27018 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction