missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein E7 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7367-ec-1mg

 View more versions of this product

Product Code. 16048480

  • £1735.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Protein E7 Protein
Research Use Only
E7
Recombinant Protein
E. coli
MHGPKATLQD IVLHLEPQNE IPVDLLCHEQ LSDSEEENDE IDGVNHQHLP ARRAEPQRHT MLCMCCKCEA RIKLVVESSA DDLRAFQQLF LNTLSFVCPW CASQQ
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers