missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein MEMO1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein MEMO1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8268-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001131074.1 | |
Protein MEMO1 Protein | |
Research Use Only | |
MEMO1 | |
Recombinant Protein | |
E. coli | |
MSNRVVCREA SHAGSWYTAS GPQLNAQLEG WLSQVQSTKR PARAIIAPHA GYTYCGSCAA HAYKQVDPSI TRRIFILGPS HHVPLSRCAL SSVDIYRTPL YDLRIDQKIY GELWKTGMFE RMSLQTDEDE HSIEMHLPYT AKAMESHKDE FTIIPVLVGA LSESKEQEFG KLFSKYLADP SNLFVVSSDF CHWGQRFRYS YYDESQGEIY RSIEHLDKMG MSIIEQLDPV SFSNYLKKYH NTICGRHPIG VLLNAITELQ KNGMNMSFSF LNYAQSSQCR NWQDSSVSYA AGALTVH | |
Greater than 80% as determined by SDS-PAGE. |
51072 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |