missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Putative inactive neutral ceramidase B Protein
A DNA sequence encoding the Homo sapiens (Human) Putative inactive neutral ceramidase B, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001072984.1 |
---|---|
Gene ID (Entrez) | 653308 |
Name | Putative inactive neutral ceramidase B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956969
|
enQuireBio™
QP5682-ec-10ug |
10 μg | |||||||
15996969
|
enQuireBio™
QP5682-ec-50ug |
50 μg | |||||||
15946969
|
enQuireBio™
QP5682-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15976969
|
enQuireBio™
QP5682-ec-200ug |
200 μg | |||||||
15986969
|
enQuireBio™
QP5682-ec-500ug |
500 μg | |||||||
15966969
|
enQuireBio™
QP5682-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001072984.1 | |
Putative inactive neutral ceramidase B Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
653308 | |
Research Use Only | |
ASAH2B | |
Recombinant Protein | |
E. coli | |
MRQHRQFMDR THYLLTFSSS ETLLRLLLRI VDRAPKGRTF GDVLQPAKPE YRVGEVAEVI FVGANPKNSV QNQTHQTFLT VEKYEATSTS WQIVCNDASW ETRFYWHKGL LGLSNATVEW HIPDTAQPGI YRIRYFGHNR KQDILKPAVI LSFEGTSPAF EVVTI | |
Greater than 90% as determined by SDS-PAGE. |