missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pyridoxal phosphate phosphatase Protein
A DNA sequence encoding the Homo sapiens (Human) Pyridoxal phosphate phosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9859-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_064711.1 | |
Pyridoxal phosphate phosphatase Protein | |
Research Use Only | |
Pdxp | |
Recombinant Protein | |
Yeast | |
MARCERLRGA ALRDVLGRAQ GVLFDCDGVL WNGERAVPGA PELLERLARA GKAALFVSNN SRRARPELAL RFARLGFGGL RAEQLFSSAL CAARLLRQRL PGPPDAPGAV FVLGGEGLRA ELRAAGLRLA GDPSAGDGAA PRVRAVLVGY DEHFSFAKLR EACAHLRDPE CLLVATDRDP WHPLSDGSRT PGTGSLAAAV ETASGRQALV VGKPSPYMFE CITENFSIDP ARTLMVGDRL ETDILFGHRC GMTTVLTLTG VSRLEEAQAY LAAGQHDLVP HYYVESIADL TEGLED | |
Greater than 90% as determined by SDS-PAGE. |
57026 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |