missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human R-Spondin 1 Protein

Product Code. 18178530
Click to view available options
Formulation:
Lyophilized from a 0.2 um filtered solution in PBS.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
1mg
25µg
This item is not returnable. View return policy

Product Code. 18178530

Brand: R&D Systems™ 4645RS01M/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is 1-10.0 ng/mL in the presence of 5 ng/mL Recombinant Mouse Wnt-3a (Catalog # 1324-WN ).

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2MKA7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in PBS.
Gene ID (Entrez) 284654
Molecular Weight (g/mol) 25.6 kDa
Name R-Spondin 1
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human R-Spondin 1 protein Ser21-Ala263
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 ug of the protein by the LAL method.
Gene Symbol RSPO1
Conjugate Unconjugated
Cross Reactivity R-Spondin 1
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 ug/mL in PBS.
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.