missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Rab GDP dissociation inhibitor beta Protein

A DNA sequence encoding the Homo sapiens (Human) Rab GDP dissociation inhibitor beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6077-ec-100ug

 View more versions of this product

Product Code. 15929809

  • £384.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001108628.1
Rab GDP dissociation inhibitor beta Protein
Research Use Only
GDI2
Recombinant Protein
E. coli
MNEEYDVIVL GTGLTECILS GIMSVNGKKV LHMDRNPYYG GESASITPLE DLYKRFKIPG SPPESMGRGR DWNVDLIPKF LMANGQLVKM LLYTEVTRYL DFKVTEGSFV YKGGKIYKVP STEAEALASS LMGLFEKRRF RKFLVYVANF DEKDPRTFEG IDPKKTTMRD VYKKFDLGQD VIDFTGHALA LYRTDDYLDQ PCYETINRIK LYSESLARYG KSPYLYPLYG LGELPQGFAR LSAIYGGTYM LNKPIEEIIV QNGKVIGVKS EGEIARCKQL ICDPSYVKDR VEKVGQVIRV ICILSHPIKN TNDANSCQII IPQNQVNRKS DIYVCMISFA HNVAAQGKYI AIVSTTVETK EPEKEIRPAL ELLEPIEQKF VSISDLLVPK DLGTESQIFI SRTYDATTHF ETTCDDIKNI YKRMTGSEFD FEEMKRKKND IYGED
Greater than 90% as determined by SDS-PAGE.
2665
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers