missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB31 / Rab22B Protein

A DNA sequence encoding the Homo sapiens (Human) RAB31 / Rab22B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7541-ec-10ug

 View more versions of this product

Product Code. 16059270

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_006859.2
RAB31 / Rab22B Protein
Research Use Only
RAB31
Recombinant Protein
E. coli
MAIRELKVCL LGDTGVGKSS IVCRFVQDHF DHNISPTIGA SFMTKTVPCG NELHKFLIWD TAGQERFHSL APMYYRGSAA AVIVYDITKQ DSFYTLKKWV KELKEHGPEN IVMAIAGNKC DLSDIREVPL KDAKEYAESI GAIVVETSAK NAINIEELFQ GISRQIPPLD PHENGNNGTI KVEKPTMQAS RRCC
Greater than 80% as determined by SDS-PAGE.
11031
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers