missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB31 / Rab22B Protein
A DNA sequence encoding the Homo sapiens (Human) RAB31 / Rab22B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7541-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_006859.2 | |
RAB31 / Rab22B Protein | |
Research Use Only | |
RAB31 | |
Recombinant Protein | |
E. coli | |
MAIRELKVCL LGDTGVGKSS IVCRFVQDHF DHNISPTIGA SFMTKTVPCG NELHKFLIWD TAGQERFHSL APMYYRGSAA AVIVYDITKQ DSFYTLKKWV KELKEHGPEN IVMAIAGNKC DLSDIREVPL KDAKEYAESI GAIVVETSAK NAINIEELFQ GISRQIPPLD PHENGNNGTI KVEKPTMQAS RRCC | |
Greater than 80% as determined by SDS-PAGE. |
11031 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |