missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB3B Protein
A cDNA sequence encoding the RAB3B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13242-20ug
This item is not returnable.
View return policy
Specifications
RAB3B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASVTDGKTG VKDASDQNFD YMFKLLIIGN SSVGKTSFLF RYADDTFTPA FVSTVGIDFK VKTVYRHEKR VKLQIWDTAG QERYRTITTA YYRGAMGFIL MYDITNEESF NAVQDWATQI KTYSWDNAQV ILVGNKCDME EERVVPTEKG QLLAEQLGFD FFEASAKENI SVRQAFERLV DAICDKMSDS LDTDPSMLGS SKNTRLSDTP PLLQQNCSC | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB3B protein 0.5 mg/ml is supplied in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 2mM DTT and 20% Glycerol. |