missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REG1B Protein
A cDNA sequence encoding the REG1B was constructed and used to recombinantly synthesize the protein.
£99.15 - £4295.00
Specifications
Name | REG1B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953109
|
enQuireBio™
QP13293-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15933109
|
enQuireBio™
QP13293-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15943109
|
enQuireBio™
QP13293-1mg |
1 mg |
£4295.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
REG1B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 20mM Tris, pH 8.0. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS HMQESQTELP NPRISCPEGT NAYRSYCYYF NEDPETWVDA DLYCQNMNSG NLVSVLTQAE GAFVASLIKE SSTDDSNVWI GLHDPKKNRR WHWSSGSLVS YKSWDTGSPS SANAGYCASL TSCSGFKKWK DESCEKKFSF VCKFKN | |
Greater than 95% as determined by SDS-PAGE. |