Learn More
R&D Systems™ Recombinant Human Relaxin R1 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7996-RR-050
Product Code. 18187130
  • £354.00 / 50µg
Explore more special offers
Description
Description
The Recombinant Human Relaxin R1 Fc Chimera Protein is derived from CHO. The Recombinant Human Relaxin R1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
69.8 kDa (monomer)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE with silver staining.
59350
Relaxin R1 Fc Chimera
Chinese Hamster Ovary cell line,CHO-derived human Relaxin R1 protein, (N-terminus) Human Relaxin R1 (Gln23-Ser398) Accession &Num; Q9HBX9 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions