missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHOD Protein
A cDNA sequence encoding the RHOD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13316-5ug
This item is not returnable.
View return policy
Specifications
RHOD Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVKVVLVGDG GCGKTSLLMV FADGAFPESY TPTVFERYMV NLQVKGKPVH LHIWDTAGQD DYDRLRPLFY PDASVLLLCF DVTSPNSFDN IFNRWYPEVN HFCKKVPIIV VGCKTDLRKD KSLVNKLRRN GLEPVTYHRG QEMARSVGAV AYLECSARLHDNVHAVFQEA AEVALSSRGR NFWRRITQGF C | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RHOD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 50% glycerol, 200mM NaCl, 2mM EDTA and 0.1mM PMSF. |