missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RING-box protein 1
A DNA sequence encoding the Homo sapiens (Human) RING-box protein 2, was expressed in the hosts and tags indicated.
Specifications
Accession Number | NP_001188299.1 |
---|---|
Gene ID (Entrez) | 9616 |
Name | RING-box protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16054540
|
enQuireBio™
QP6613-ec-10ug |
10 μg |
N/A
|
N/A | |||||
16094540
|
enQuireBio™
QP6613-ec-50ug |
50 μg |
N/A
|
N/A | |||||
16044540
|
enQuireBio™
QP6613-ec-100ug |
100 μg |
N/A
|
N/A | |||||
16074540
|
enQuireBio™
QP6613-ec-200ug |
200 μg |
N/A
|
N/A | |||||
16084540
|
enQuireBio™
QP6613-ec-500ug |
500 μg |
N/A
|
N/A | |||||
16064540
|
enQuireBio™
QP6613-ec-1mg |
1 mg |
N/A
|
N/A | |||||
Specifications
NP_001188299.1 | |
RING-box protein 1 | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
9616 | |
Research Use Only | |
RNF7 | |
Recombinant Protein | |
ADVEDGEETC ALASHSGSSG SKSGGDKMFS LKKWNAVAMW SWDVECDTCA ICRVQVMDAC LRCQAENKQE DCVVVWGECN HSFHNCCMSL WVKQNNRCPL CQQDWVVQRI GK |