missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RING finger protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) RING finger protein 11, was expressed in the hosts and tags indicated.
Specifications
Accession Number | NP_055187.1 |
---|---|
Gene ID (Entrez) | 26994 |
Name | RING finger protein 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078191
|
enQuireBio™
QP9370-ye-10ug |
10 μg |
N/A
|
N/A | |||||
16018201
|
enQuireBio™
QP9370-ye-50ug |
50 μg |
N/A
|
N/A | |||||
16068191
|
enQuireBio™
QP9370-ye-100ug |
100 μg |
N/A
|
N/A | |||||
16098191
|
enQuireBio™
QP9370-ye-200ug |
200 μg |
N/A
|
N/A | |||||
16008201
|
enQuireBio™
QP9370-ye-500ug |
500 μg |
N/A
|
N/A | |||||
16088191
|
enQuireBio™
QP9370-ye-1mg |
1 mg |
N/A
|
N/A | |||||
Specifications
NP_055187.1 | |
RING finger protein 11 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
26994 | |
Research Use Only | |
RNF11 | |
Recombinant Protein | |
Yeast | |
GNCLKSPTSD DISLLHESQS DRASFGEGTE PDQEPPPPYQ EQVPVPVYHP TPSQTRLATQ LTEEEQIRIA QRIGLIQHLP KGVYDPGRDG SEKKIRECVI CMMDFVYGDP IRFLPCMHIY HLDCIDDWLM RSFTCPSCME PVDAALLSSY ETN | |
Greater than 90% as determined by SDS-PAGE. |