missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human RPGR GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00006103-Q01-25ug
501.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification | |
6103 | |
25 μg | |
COD1, cone dystrophy 1 (X-linked), CORDX1RP3retinitis pigmentosa 15, CRD, orf15, PCDX, retinitis pigmentosa 3 GTPase regulator, retinitis pigmentosa GTPase regulator, RP15, X-linked retinitis pigmentosa GTPase regulator, XLRP3 | |
Neuroscience | |
RPGR |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
36.63 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
RPGR | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human RPGR GST (N-Term) Protein > 25 μg
Spot an opportunity for improvement?Share a Content Correction