missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPL26L1 Protein
A cDNA sequence encoding the RPL26L1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13346-100ug
This item is not returnable.
View return policy
Specifications
51121 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKFNPFV TSDRSKNRKR HFNAPSHVRR KIMSSPLSKE LRQKYNVRSM PIRKDDEVQV VRGHYKGQQI GKVVQVYRKK YVIYIERVQR EKANGTTVHV GIHPSKVVIT RLKLDKDRKK ILERKAKSRQ VGKEKGKYKE ELIEKMQE | |
Greater than 90% as determined by SDS-PAGE. |
RPL26L1 Protein | |
Research Use Only | |
RPL26L1 | |
Human | |
His | |
The RPL26L1 solutioncontains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 40% glycerol. |