missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RXRA Protein
A cDNA sequence encoding the RXRA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13378-1mg
This item is not returnable.
View return policy
Specifications
RXRA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MLGLNGVLKV PAHPSGNMAS FTKHICAICG DRSSGKHYGV YSCEGCKGFF KRTVRKDLTY TCRDNKDCLI DKRQRNRCQY CRYQKCLAMG MKREAVQEER QRGKDRNENE VESTSSANE | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) containing 20mM Tris-HCl pH 7.5, 0.1M NaCl and 5mM ?-Mercaptoethanol. |