missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SDF2 Protein
A DNA sequence encoding the Homo sapiens (Human) SDF2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_008854.2 |
---|---|
Gene ID (Entrez) | 6388 |
Name | SDF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094860
|
enQuireBio™
QP6657-ec-10ug |
10 μg | |||||||
16034870
|
enQuireBio™
QP6657-ec-50ug |
50 μg | |||||||
16084860
|
enQuireBio™
QP6657-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16014870
|
enQuireBio™
QP6657-ec-200ug |
200 μg | |||||||
16024870
|
enQuireBio™
QP6657-ec-500ug |
500 μg | |||||||
16004870
|
enQuireBio™
QP6657-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_008854.2 | |
SDF2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6388 | |
Research Use Only | |
SDF2 | |
Recombinant Protein | |
E. coli | |
SSLGVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKSAT VCERGTPIKC GQPIRLTHVN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTEVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLKAEAHH AEL | |
Greater than 90% as determined by SDS-PAGE. |