missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine racemase Protein
A DNA sequence encoding the Homo sapiens (Human) Serine racemase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6729-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_068766.1 | |
Serine racemase Protein | |
Research Use Only | |
SRR | |
Recombinant Protein | |
E. coli | |
MCAQYCISFA DVEKAHINIR DSIHLTPVLT SSILNQLTGR NLFFKCELFQ KTGSFKIRGA LNAVRSLVPD ALERKPKAVV THSSGNHGQA LTYAAKLEGI PAYIVVPQTA PDCKKLAIQA YGASIVYCEP SDESRENVAK RVTEETEGIM VHPNQEPAVI AGQGTIALEV LNQVPLVDAL VVPVGGGGML AGIAITVKAL KPSVKVYAAE PSNADDCYQS KLKGKLMPNL YPPETIADGV KSSIGLNTWP IIRDLVDDIF TVTEDEIKCA TQLVWERMKL LIEPTAGVGV AAVLSQHFQT VSPEVKNICI VLSGGNVDLT SSITWVKQAE RPASYQSVSV | |
Greater than 90% as determined by SDS-PAGE. |
63826 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |