missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal-regulatory protein beta-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein beta-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6685-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065090

  • £1405.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001077379.1
Signal-regulatory protein beta-1 Protein
Research Use Only
SIRPB1
Recombinant Protein
Mammal
EDELQVIQPE KSVSVAAGES ATLRCAMTSL IPVGPIMWFR GAGAGRELIY NQKEGHFPRV TTVSELTKRN NLDFSISISN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAVRA TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP AGDSVSYSIH STARVVLTRG DVHSQVICEI AHITLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRAENQAN VTCQVSNFYP RGLQLTWLEN GNVSRTETAS TLIENKDGTY NWMSWLLVNT CAHRDDVVLT CQVEHDGQQA VSKSYALEIS AHQKEHGSDI THEAALAPTA PL
Greater than 90% as determined by SDS-PAGE.
10326
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers