missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIRT1 / SIR2L1 Protein
A DNA sequence encoding the Homo sapiens (Human) SIRT1 / SIR2L1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001135970.1 |
---|---|
Gene ID (Entrez) | 23411 |
Name | SIRT1 / SIR2L1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943799
|
enQuireBio™
QP1351-ec-10ug |
10 μg | |||||||
15983799
|
enQuireBio™
QP1351-ec-50ug |
50 μg | |||||||
15933799
|
enQuireBio™
QP1351-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963799
|
enQuireBio™
QP1351-ec-200ug |
200 μg | |||||||
15973799
|
enQuireBio™
QP1351-ec-500ug |
500 μg | |||||||
15953799
|
enQuireBio™
QP1351-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001135970.1 | |
SIRT1 / SIR2L1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
23411 | |
Research Use Only | |
SIRT1 | |
Recombinant Protein | |
E. coli | |
ADEAALALQP GGSPSAAGAD REAASSPAGE PLRKRPRRDG PGLERSPGEP GGAAPEREVP AAARGCPGAA AAALWREAEA EAAAAGGEQE AQATAAAGEG DNGPGLQGPS REPPLADNLY DEDDDDEGEE EEEAAAAAIG YRDNLLFGDE IITNGFHSCE SDEEDRASHA SSSDWTPRPR IGPYTFVQQH LMIGTDPRTI LKDLLPETIP PPELDDMTLW QIVINILSEP PKRKKRKDIN TIEDAVKLLQ ECKKIIVLTG AGVSVSCGIP DFRSRDGIYA RLAVDFPDLP DPQAMFDIEY FRKDPRPFFK FAKEIYPGQF QPSLCHKFIA LSDKEGKLLR NYTQNIDTLE QVAGIQRIIQ CHGSFATASC LICKYKVDCE AVRGDIFNQV VPRCPRCPAD EPLAIMKPEI VFFGENLPEQ FHRAMKYDKD EVDLLIVIGS SLKVRPVALI PSSIPHEVPQ ILINREPLPH LHFDVELLGD CDVIINELCH RLGGEYAKLC CNPVKLSEIT EKPPRTQKEL AYLSELPPTP LHVSEDSSSP ERTSPPDSSV IVTLLDQAAK SNDDLDVSES KGCMEEKPQE VQTSRNVESI AEQMENPDLK NVGSSTGEKN ERTSVAGTVR KCWPNRVAKE QISRRLDGNQ YLFLPPNRYI FHGAEVYSDS EDDVLSSSSC GSNSDSGTCQ SPSLEEPMED ESEIEEFYNG LEDEPDVPER AGGAGFGTDG DDQEAINEAI SVKQEVTDMN YPSNKS | |
Greater than 90% as determined by SDS-PAGE. |